Learn More
Sigma Aldrich Fine Chemicals Biosciences Low Profile Roller Lab Sta1EA
Catalog No. 502909000
Encompass
The IBI Low Profile Roller is a lightweight unit designed as a compact space saving non-dedicated instrument. This unit will operate on a benchtop inside high humidity and CO2incubators high temperature ovens (60C) and refrigerators (0C). The IBI Low Profile Roller comes standard with six rollers in place for rotating three large cell culture bottles. The versatility of this unit allows the user to add up to five additional rollers quickly and easily. No tools are needed to move or add rollers. Moving rollers closer together or adding more rollers allows this system to roll small tubes from 15mL to 50mL. Rubber O-Rings to be placed on the small tubes are included with every unit.The low profile design provides a convenient fit into laboratory incubators ovens and refrigerated units. The thin flat 115V power cord fits easily through incubator doors without compromising the environment inside. An industrial grade chain drive provides years of trouble-free service. All product fin
Catalog No. 50-290-9000 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7420011EA
The IBI Low Profile Roller is a lightweight unit designed as a compact space saving non-dedicated instrument. This unit will operate on a benchtop inside high humidity and CO2incubators high temperature ovens (60C) and refrigerators (0C). The IBI Low Profile Roller comes standard with six rollers in place for rotating three large cell culture bottles. The versatility of this unit allows the user to add up to five additional rollers quickly and easily. No tools are needed to move or add rollers. Moving rollers closer together or adding more rollers allows this system to roll small tubes from 15mL to 50mL. Rubber O-Rings to be placed on the small tubes are included with every unit.The low profile design provides a convenient fit into laboratory incubators ovens and refrigerated units. The thin flat 115V power cord fits easily through incubator doors without compromising the environment inside. An industrial grade chain drive provides years of trouble-free service. All product fin