Learn More
Sigma Aldrich Fine Chemicals Biosciences Low profile roller low pro1EA
Catalog No. 502908843
Encompass
Weather-proof roller operates on benchtop in dry or humid incubators in cold rooms to 0C and even in ovens to 65C. Use for cell culture with standard roller bottles and in filter blot applications with hybridization bottles (below). Unit is supplied with six gear-driven rollers to rotate three large bottles at variable speed 0.25-8 rpm. Rollers can be moved closer together to accommodate small bottles and tubes and up to five more rollers can be added as many as thirty 10AxA75Amm tubes can be turned at 2-67Arpm. Unit is powered by a cool-running 24AVDC motor. Thin flat AC power cord (US only) reaches external outlet without damaging incubator or refrigerator gasket.
Catalog No. 50-290-8843 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3677291EA
Weather-proof roller operates on benchtop in dry or humid incubators in cold rooms to 0C and even in ovens to 65C. Use for cell culture with standard roller bottles and in filter blot applications with hybridization bottles (below). Unit is supplied with six gear-driven rollers to rotate three large bottles at variable speed 0.25-8 rpm. Rollers can be moved closer together to accommodate small bottles and tubes and up to five more rollers can be added as many as thirty 10AxA75Amm tubes can be turned at 2-67Arpm. Unit is powered by a cool-running 24AVDC motor. Thin flat AC power cord (US only) reaches external outlet without damaging incubator or refrigerator gasket.