Learn More
CHEMetrics™ by AquaPhoenix Low Range Nitrite Comparator

Description
- C-7006 Nitrite Low Range Comparator is a component of K-7006 Nitrite CHEMets Visual Test Kit and is designed to be compared against the color formed in R-7006 Nitrite CHEMets Refill.
- The comparator's range is applicable to many industrial and environmental applications. Color standards are pink in color and cover a range of 0 to 0.10 mg/L nitrite (as nitrogen).
Specifications
Specifications
| pH Range | 7 |
| Volume (Metric) | 0.5 mL |
| For Use With (Equipment) | K-7006 |
| Type | Nitrite Comparator |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.