Learn More
INDUSTRIAL TEST SYSTEMS INC Peroxide Test Strip, Low Range, 50 Seconds Test Time, 0.05-4 ppm Range (Bottle of 50)
SDP
Catalog No. NC0104002
Encompass_Preferred
This item has a minimum qty of 10 per supplier requirements.
Combining accuracy, affordability, and ease-of-use, our patented test strips have revolutionized water testing. They provide accurate, rapid results at an affordable cost. Designed using cutting-edge chemistries and state-of-the-art manufacturing processes, our test strips and test kits replace outdated testing kits that are labor and cost intensive.
Catalog No. NC0104002 Supplier INDUSTRIAL TEST SYSTEMS INC Supplier No. 481015
This item has a minimum qty of 10 per supplier requirements.
Combining accuracy, affordability, and ease-of-use, our patented test strips have revolutionized water testing. They provide accurate, rapid results at an affordable cost. Designed using cutting-edge chemistries and state-of-the-art manufacturing processes, our test strips and test kits replace outdated testing kits that are labor and cost intensive.