Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Low Range Phosphate, Ortho Comparator
SDP

Catalog No. 13299647
Change view
Click to view available options
For Use With (Equipment):
K-8510
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
13-299-647 K-8510
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-647 Supplier CHEMetrics™ by AquaPhoenix Supplier No. C8501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Low range ortho phosphate comparator designed for comparison against color formed during the reaction. Supports visual analysis of phosphate test results.

  • C-8501 Low Range Phosphate, ortho Comparator is a component of K-8510 Phosphate,ortho CHEMets Visual Test Kit and is designed to be compared against the color formed in R-8510 Phosphate, ortho CHEMets Refill.
  • The comparator's range is applicable to many environmental and water treatment applications. Color standards are blue in color and cover a range of 0-1.0 ppm PO4.

Specifications

pH Range 7
Volume (Metric) 0.5 mL
For Use With (Equipment) K-8510
Range 0 to 1 ppm
Type Phosphate Comparator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.