Learn More
Research Products International Corp Low Retention Tips, 1-200ul, Natural Color, Non-Sterile, Bulk, 1000 per case
SDP
Catalog No. 502114065
Encompass
Low Retention Pipet Tips offer complete sample recovery, eliminate sample retention and increase pipetting accuracy. Ideal tip for minimum sample and reagent retention. Graduated, super clear tips feature accurate measurements, universal airtight fit, and are perfect for highly sensitive PCE and microbiology applications. High performance tips are certified free of detectable Human DNA, RNase, DNase, pyrogens & PCR Inhibitors.
Catalog No. 50-211-4065 Supplier Research Products International Corp Supplier No. 246238GL
Low Retention Pipet Tips offer complete sample recovery, eliminate sample retention and increase pipetting accuracy. Ideal tip for minimum sample and reagent retention. Graduated, super clear tips feature accurate measurements, universal airtight fit, and are perfect for highly sensitive PCE and microbiology applications. High performance tips are certified free of detectable Human DNA, RNase, DNase, pyrogens & PCR Inhibitors.