Learn More
Midwest Scientific PR1MA Low Retention Graduated Barrier Pipette Tips, 10ul, 10 racks of 96 tips
SDP
Catalog No. NC1959255
Encompass
PR1MA™ Pipette Tips were meticulously designed using a specially formulated NoStick resin to provide researchers with the highest level of liquid handling performance. PR1MA™ Pipette Tips were created with the belief that when it comes to liquid handling, less is more.Less Fluid Retention. Less Sample Loss. Less Time Spent. Less Plastic Wasted. Filtered tips have a hydrophobic HDPE barrier that won't contaminate or impede airflow.PR1MA™ Pipette Tips are produced under strict controls and an ISO9001 certified Quality Assurance process. They are manufactured with a state of the art fully automated tip manufacturing, filtering and inspecting which is incomparable in our industry.
Catalog No. NC1959255 Supplier Midwest Scientific Supplier No. PR10RKFL
PR1MA™ Pipette Tips were meticulously designed using a specially formulated NoStick resin to provide researchers with the highest level of liquid handling performance. PR1MA™ Pipette Tips were created with the belief that when it comes to liquid handling, less is more.Less Fluid Retention. Less Sample Loss. Less Time Spent. Less Plastic Wasted. Filtered tips have a hydrophobic HDPE barrier that won't contaminate or impede airflow.PR1MA™ Pipette Tips are produced under strict controls and an ISO9001 certified Quality Assurance process. They are manufactured with a state of the art fully automated tip manufacturing, filtering and inspecting which is incomparable in our industry.