Learn More
Sigma Aldrich Fine Chemicals Biosciences ART(R) low retention SoftFit L Reload System(TM) maximum volume 1000 muL, Tip design ART 1000L, Hinged rack | 4X768EA
Catalog No. NC1965417
Encompass
ART(R) low retention SoftFit L Reload System(TM) maximum volume 1000 muL, Tip design ART 1000L, Hinged rack | 4X768EA
Catalog No. NC1965417 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6770194X768EA
ART(R) low retention SoftFit L Reload System(TM) maximum volume 1000 muL, Tip design ART 1000L, Hinged rack | 4X768EA