Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Low Retention Universal Fit Pipette Tip, Clear, Nonsterile

Catalog No. 01677292 Shop All Axygen Products
Change view
Click to view available options
Format:
Bulk
Rack
Tip Style:
Beveled
Fine Point
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Tip Style
01-677-292 Rack Beveled
01-677-271 Bulk Beveled
01-677-226 Rack Fine Point
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-677-292 Supplier Corning™ Supplier No. P30102372

missing translation for 'validMainframeMsg'

RNase-/DNase-free

  • Universal fit pipette tips deliver improved pipettor compatibility durable hinged racks and a complete offering from 10 μL to 1000 μL
  • Features include extended length tips wide bore designs for viscous solutions and low retention technology to enhance liquid handling performance
  • RNaseDNase free
  • Nonpyrogenic
  • Human DNA free
  • ATP free
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Rack
Tip Style Beveled
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.