Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Low Serum Growth Supplement (LSGS)

Catalog No. S00310
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S00310 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S00310 Supplier Gibco™ Supplier No. S00310
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Low Serum Growth Supplement (LSGS)

Low Serum Growth Supplement (LSGS) is a sterile, concentrated (50X) solution intended for use as one component in a complete culture environment for the growth of human dermal fibroblasts and endothelial cells derived from large vessels. LSGS is intended for use with Medium 106 (fibroblasts) or Medium 200 (endothelial cells). LSGS contains: fetal bovine serum, basic fibroblast growth factor, heparin, hydrocortisone, and epidermal growth factor. When a 500 ml bottle of medium is supplemented with LSGS or the components of the LSGS Kit, the final concentrations of the components in the supplemented medium are:
  • fetal bovine serum, 2% v/v
  • hydrocortisone (1μg/ml)
  • human epidermal growth factor, 10 ng/ml
  • basic fibroblast growth factor, 3 ng/ml
  • heparin, 10μg/ml
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store in freezer (-5 to -20°C).
    Cell Type Epithelial Cells (Large Vessel), Fibroblast Cells (Dermal)
    Concentration 50 X
    Product Type Growth Supplement
    Sterility Sterile
    Product Line Cascade Biologics
    Quantity 10 mL
    Shipping Condition Dry Ice
    Species Human
    WARNING: Cancer - www.P65Warnings.ca.gov

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.