Learn More
GRANT INSTRUMENTS Centrifuge low speed benchtop, 0 - 3000 rpm, 12 x 2-50ml tubes/2 x plates. Order rotor separately
SDP
Catalog No. NC2176888
Encompass_Preferred
The Grant LMC-3000 is a low speed benchtop centrifuge for biomedical applications.It has interchangeable rotors for accommodating centrifuge tubes (10 to 15 ml or 50 ml) or standard 96-well microtitre plates.Suitable for a wide range of analytical applications including biomedical, bio-organic and immunoenzyme analysis.Spin speed: up to 3000 rpm for tubes, up to 2000 rpm for microtitre plates.Timed operation (1 to 90 minutes), with automatic switch-off.Choice of interchangeable rotors for up to 12 centrifuge tubes or 2 microtitre plates.Easy set-up of spin speed and time via 2-line LCD display and simple push buttons.Sturdy metal housing and lid, automatic imbalance switch-off and lid lock when centrifuge is running provide for safe operation in your workplace.Compact design with small footprint - fits neatly into the workspace.
This item is not returnable. View return policy
Catalog No. NC2176888 Supplier GRANT INSTRUMENTS Supplier No. LMC3000
The Grant LMC-3000 is a low speed benchtop centrifuge for biomedical applications.It has interchangeable rotors for accommodating centrifuge tubes (10 to 15 ml or 50 ml) or standard 96-well microtitre plates.Suitable for a wide range of analytical applications including biomedical, bio-organic and immunoenzyme analysis.Spin speed: up to 3000 rpm for tubes, up to 2000 rpm for microtitre plates.Timed operation (1 to 90 minutes), with automatic switch-off.Choice of interchangeable rotors for up to 12 centrifuge tubes or 2 microtitre plates.Easy set-up of spin speed and time via 2-line LCD display and simple push buttons.Sturdy metal housing and lid, automatic imbalance switch-off and lid lock when centrifuge is running provide for safe operation in your workplace.Compact design with small footprint - fits neatly into the workspace.