Learn More
Malvern Panalytical Low Sulfur 33Mm Sample Cups
Catalog No. NC1363833
Encompass
One case of 1152 preassembled 35 mm cups. Each cup is preassembled with 89 mm diameter 3.5 micron low sulfur mylar film and is ready for use. The cups are packaged in specially designed trays containing twelve cups. Each box contains 4 trays of cups and each case contains 24 boxes.
Catalog No. NC1363833 Supplier Malvern Panalytical Supplier No. 10048214
One case of 1152 preassembled 35 mm cups. Each cup is preassembled with 89 mm diameter 3.5 micron low sulfur mylar film and is ready for use. The cups are packaged in specially designed trays containing twelve cups. Each box contains 4 trays of cups and each case contains 24 boxes.