Learn More
Electron Microscopy Sciences Low Temperature Desiccator Cabinet, Bronze
SDP
Catalog No. 50930914
Encompass_Preferred
Dry, dust-free storage for use in refrigerators, freezers or at room temperature. These durable cabinets are made from 6.35 mm ( 1/4") acrylic with full-length hinges and stainless steel twist locking latches. Deep slide-in trays have perforated bottoms.
Footprint: 9" x 9"
Height: 16"
Catalog No. 50-930-914 Supplier Electron Microscopy Sciences Supplier No. 71220A
Dry, dust-free storage for use in refrigerators, freezers or at room temperature. These durable cabinets are made from 6.35 mm ( 1/4") acrylic with full-length hinges and stainless steel twist locking latches. Deep slide-in trays have perforated bottoms.
Footprint: 9" x 9"
Height: 16"