Learn More
Electron Microscopy Sciences Low-Temp Hot Glue Gun
SDP
Catalog No. 5026986
Encompass_Preferred
This low temperature mini glue gun is safe and allows for the easy application of the melt glue to the aluminum stub for the mounting of bulk or irregular shaped specimens for SEM. Comes complete with a 40" cord, and two glue sticks. Accepts 5/16" diameter all temperature or low temperature glue sticks. 110 volt, 10 watts.
Catalog No. 50-269-86 Supplier Electron Microscopy Sciences Supplier No. 7217100
This low temperature mini glue gun is safe and allows for the easy application of the melt glue to the aluminum stub for the mounting of bulk or irregular shaped specimens for SEM. Comes complete with a 40" cord, and two glue sticks. Accepts 5/16" diameter all temperature or low temperature glue sticks. 110 volt, 10 watts.