Learn More
Yamato Scientific America, Inc. Yamato 143L Refrigerant Incubator with window and MK161 shaker, 115V
Catalog No. 501898106 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Low temperature 143L forced convection incubator applicable for low temperature tests and environmental tests. Designed with a large dual glass door and inner door that forms a triplex glass door for improved heat retention. Programmable. High accuracy temperature control and temperature distribution. Optional slide shaker table available to put in and take out samples easily. Comes with a compact shaker. Operating temperature range: -10~ + 50° C
Catalog No. 50-189-8106 Supplier Yamato Scientific America, Inc. Supplier No. IN604WMK 115V
Add to Cart
Add to Cart
Request a Quote
Low temperature 143L forced convection incubator applicable for low temperature tests and environmental tests. Designed with a large dual glass door and inner door that forms a triplex glass door for improved heat retention. Programmable. High accuracy temperature control and temperature distribution. Optional slide shaker table available to put in and take out samples easily. Comes with a compact shaker. Operating temperature range: -10~ + 50° C