Learn More
Yamato Scientific America, Inc. Yamato 286L Incubator, Refrigerant, 220V
Catalog No. 501898101 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Low temperature 286L forced convection incubator applicable for low temperature tests and environmental tests. Programmable. High accuracy temperature control and temperature distribution. Inner glass door keeps temperature stable during sample observation. Operating temperature range: -10~ + 50° C
Catalog No. 50-189-8101 Supplier Yamato Scientific America, Inc. Supplier No. IN804 220V
Add to Cart
Add to Cart
Request a Quote
Low temperature 286L forced convection incubator applicable for low temperature tests and environmental tests. Programmable. High accuracy temperature control and temperature distribution. Inner glass door keeps temperature stable during sample observation. Operating temperature range: -10~ + 50° C