Learn More
Yamato Scientific America, Inc. Yamato 143L Incubator with window, Refrigerant, 115V
Catalog No. 501898102 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Low temperature 143L forced convection incubator applicable for low temperature tests and environmental tests. Designed with a large dual glass door and inner door that forms a triplex glass door for improved heat retention. Programmable. High accuracy temperature control and temperature distribution. Optional slide shaker table available to put in and take out samples easily. Operating temperature range: -10~ + 50° C
Catalog No. 50-189-8102 Supplier Yamato Scientific America, Inc. Supplier No. IN604W 115V
Add to Cart
Add to Cart
Request a Quote
Low temperature 143L forced convection incubator applicable for low temperature tests and environmental tests. Designed with a large dual glass door and inner door that forms a triplex glass door for improved heat retention. Programmable. High accuracy temperature control and temperature distribution. Optional slide shaker table available to put in and take out samples easily. Operating temperature range: -10~ + 50° C