Learn More
Percival Scientific Low Temperature Chamber, LT105C9, 122.9 cu. ft., SciWhite with red LEDs, 1 tier, -10 - 44° C ±0.5°, dual coil refrigeration, IntellusUltraConnect controller, touchscreen
SDP
Catalog No. NC3795641
Encompass_Preferred
The LT105C9 chamber is used for cold hardiness and exposure to multi temperatures.Vertically adjustable lamp bank of SciWhite w/ red LEDs dimmable from 10-100% output w/ up to 1450 µmoles/m2/s irradiance @ 6". IntellusUltra control system w/ touchscreen interface, remote connectivity. Constant temperature defrost allows chamber to operate at -10-44°C ±0.5° lights on w/ dual coil refrigeration system.Conditioned air moves in upward direction through work bench through perforations in aluminum channels. Interior/exterior of smooth white galvanized steel w/ foamed-in-place non-CFC insulation and stainless-steel floor for corrosion resistance. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; also prolongs life of compressor, and eliminates risk of ice build up in coil For additional options contact sales@percival-scientific.com.
Catalog No. NC3795641 Supplier Percival Scientific Supplier No. LT105C9
Add to Cart
Add to Cart
Request a Quote
The LT105C9 chamber is used for cold hardiness and exposure to multi temperatures.Vertically adjustable lamp bank of SciWhite w/ red LEDs dimmable from 10-100% output w/ up to 1450 µmoles/m2/s irradiance @ 6". IntellusUltra control system w/ touchscreen interface, remote connectivity. Constant temperature defrost allows chamber to operate at -10-44°C ±0.5° lights on w/ dual coil refrigeration system.Conditioned air moves in upward direction through work bench through perforations in aluminum channels. Interior/exterior of smooth white galvanized steel w/ foamed-in-place non-CFC insulation and stainless-steel floor for corrosion resistance. Self-contained air-cooled condensing unit with hot gas bypass system for continuous compressor operation extends life, ensuring precise temperature control by alternately cycling refrigerant and hot gas to coil; also prolongs life of compressor, and eliminates risk of ice build up in coil For additional options contact sales@percival-scientific.com.