Learn More
Andwin Scientific BDH Aristar Plus Water HPLC LOW TOC grade, 4x4L
SDP
Catalog No. NC0674433
Encompass_Preferred
The water with low TOC can be used for analytical and preparative separations, gradient and isocratic conditions, and LC, HPLC, and trace organic analysis. This product is packaged in a 4-liter amber glass bottle and is sold in a case of 4.
Catalog No. NC0674433 Supplier Andwin Scientific Supplier No. 87003652CS
The water with low TOC can be used for analytical and preparative separations, gradient and isocratic conditions, and LC, HPLC, and trace organic analysis. This product is packaged in a 4-liter amber glass bottle and is sold in a case of 4.