Learn More
Cederlane Low-Tox Guinea Pig Complement, lyophilized, 5x1 mL, low toxicity to mouse lymphocytes
SDP
Catalog No. NC1308691
Encompass
The CEDARLANE Low-Tox Guinea Pig Complement is a lyophilized reagent designed for high complement activity while minimizing toxicity for sensitive immunological assays. Sourced from guinea pigs, it is absorbed to lower toxicity, particularly to mouse lymphocytes, making it ideal for cytotoxicity and plaque assays without additional processing. The lyophilized format provides stability and ease of storage, ensuring high performance in various applications.
  • Processed to ensure high complement activity
  • Minimal toxicity to mouse lymphocytes
  • Absorbed to eliminate background toxicity
  • Suitable for cytotoxicity assays
  • Suitable for plaque assays
  • Storage stability at -20°C (lyophilized)
  • Reconstitutable with 1 mL or 5 mL of ice cold distilled water
  • Shipping requires blue ice
  • High complement activity
Catalog No. NC1308691 Supplier Cederlane Supplier No. CL4051
The CEDARLANE Low-Tox Guinea Pig Complement is a lyophilized reagent designed for high complement activity while minimizing toxicity for sensitive immunological assays. Sourced from guinea pigs, it is absorbed to lower toxicity, particularly to mouse lymphocytes, making it ideal for cytotoxicity and plaque assays without additional processing. The lyophilized format provides stability and ease of storage, ensuring high performance in various applications.
  • Processed to ensure high complement activity
  • Minimal toxicity to mouse lymphocytes
  • Absorbed to eliminate background toxicity
  • Suitable for cytotoxicity assays
  • Suitable for plaque assays
  • Storage stability at -20°C (lyophilized)
  • Reconstitutable with 1 mL or 5 mL of ice cold distilled water
  • Shipping requires blue ice
  • High complement activity