Learn More
Electron Microscopy Sciences Low viscosity embedding media kit, 1 kit
SDP
Catalog No. 50980409
Encompass_Preferred
Provides excellent penetration for embedding biological tissues and rapid infiltration. Easy to prepare. The hardness is adjusted by changing the proportion of the flexibilizer (DER 736). The blocks have good trimming and sectioning qualities. Thin sections are tough under the electron beam. Kit consists of: 450 ml NSA, 225 ml ERL 4221 , 225 ml DER 736 , 25 ml DMAE.
Catalog No. 50-980-409 Supplier Electron Microscopy Sciences Supplier No. 14300
Provides excellent penetration for embedding biological tissues and rapid infiltration. Easy to prepare. The hardness is adjusted by changing the proportion of the flexibilizer (DER 736). The blocks have good trimming and sectioning qualities. Thin sections are tough under the electron beam. Kit consists of: 450 ml NSA, 225 ml ERL 4221 , 225 ml DER 736 , 25 ml DMAE.