Learn More
Grainger Low Volt NP Digital Tstat H or C Wired
Catalog No. 502768139
Encompass_Preferred
The Honeywell RTH6580WF 7-Day Programmable Thermostat is a low voltage, programmable thermostat that allows you to set customized heating and cooling schedules for 7 days. It has a touchscreen interface and is compatible with conventional and heat pump systems. The thermostat can be programmed for 4 time periods per day and has an adaptive intelligent recovery feature that learns patterns and adjusts accordingly. It requires 2 AA batteries for backup power and can control 1 heat/1 cool system. The thermostat has a slim, wall-hugging design.
Catalog No. 50-276-8139 Supplier Grainger Supplier No. 16X609
The Honeywell RTH6580WF 7-Day Programmable Thermostat is a low voltage, programmable thermostat that allows you to set customized heating and cooling schedules for 7 days. It has a touchscreen interface and is compatible with conventional and heat pump systems. The thermostat can be programmed for 4 time periods per day and has an adaptive intelligent recovery feature that learns patterns and adjusts accordingly. It requires 2 AA batteries for backup power and can control 1 heat/1 cool system. The thermostat has a slim, wall-hugging design.