Learn More
Electron Microscopy Sciences SEM Low Voltage Resolution Specimen Gold on Carbon, Hitachi Stub
SDP
Supplier:  Electron Microscopy Sciences 7951304
Encompass_Preferred
Larger gold particles in this test allow for a higher contrast and smaller gaps allow resolution to be measured. This helps to make the test easier to use when operating conditions are not optimal.
Type: Hitachi Stub
Catalog No. 50-193-1208