Learn More
Corning™ Low Volume 384-well Black/Clear Flat Bottom Polystyrene Poly-D-Lysine Coated Microplate with Lid, Aseptically Manufactured
Catalog No. 07202470 Shop All Corning Products
Change view
Click to view available options
Color:
Black, Clear Bottom
Catalog No. Color
07-202-470 Black, Clear Bottom
1 options
Catalog No. 07-202-470 Supplier Corning™ Supplier No. 3643
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Ideal for miniaturized 384 well fluorescent cell-based assays
  • Round well design with a clear flat bottom
  • 2.8 mm distance between well bottoms and plate bottom
  • 50 μL total well volume, with recommended working volume of 5-40 μL
  • High base plate design is compatible with automation and for bar coding
  • Validated by Molecular Devices for the FLIPR reader. Outer wells aligned for proper viewing with FLIPR
  • Poly-D-Lysine coated for enhanced cell attachment
  • Sterile (aseptic assembly) and certified nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Color Black, Clear Bottom
Material Polystyrene
No. of Wells 384
Volume (Metric) Working 5 to 40 μL
Sterility Sterile
Surface Treatment Poly-D-Lysine
Well Shape Flat Bottom (F-well)
Quantity 10/Pk., 50/Cs.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov