Learn More
Poltex Low Volume Elution Riser for MAGBEAD520
Description
- Riser unit to allow for low volume magnetic bead extraction
- Brings bottom of microtube higher up toward magnet when solution volume is low
- Works with the Poltex MAGBEAD magnetic bead separation racks (sold separately)
Specifications
Specifications
| Product Type | Riser |
| No. of Doors | None |
| Color | Clear |
| Autoclavable | Not Autoclavable |
| No. of Shelves | 1 |
| Material | Acrylic |