Learn More
Hach Company Low-volume flow chamber
Catalog No. 5000207123 Shop All Hach Company Products
Encompass
Low-volume flow chamber-PVDF-3422-series sensorsFor use with the 3422 Contacting Conductivity Sensors K 0 05 Cell Constant Only PVDF 1/2-Inch Pipe Tee Limits sample volume to approximately 20 ml for high-purity water applications
Catalog No. 50-002-07123 Supplier Hach Company Supplier No. 1000G3316101
Low-volume flow chamber-PVDF-3422-series sensorsFor use with the 3422 Contacting Conductivity Sensors K 0 05 Cell Constant Only PVDF 1/2-Inch Pipe Tee Limits sample volume to approximately 20 ml for high-purity water applications