Learn More
Waters Corp Waters Low Volume Insert, Clear Glass, 250 μL, 4 mL Vial Compatible
Catalog No. 50785072 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Vial inserts are used in applications with limited sample volume. This clear glass low volume insert has a 250 μL capacity and is designed for use with 4 mL screw cap vials.
  • Designed for applications with limited sample volume.
  • Has a 250 μL capacity.
  • Made from clear glass.
  • Compatible with 4 mL screw cap vials.
  • Packaged with 144 units per package.
  • Requires a spring for centering within the vial (sold separately).
Catalog No. 50-785-072 Supplier Waters Corp Supplier No. WAT015199
Vial inserts are used in applications with limited sample volume. This clear glass low volume insert has a 250 μL capacity and is designed for use with 4 mL screw cap vials.
  • Designed for applications with limited sample volume.
  • Has a 250 μL capacity.
  • Made from clear glass.
  • Compatible with 4 mL screw cap vials.
  • Packaged with 144 units per package.
  • Requires a spring for centering within the vial (sold separately).