Learn More
Waters Corp Waters Glass Insert, 250 μL, Glass
Catalog No. 50785887 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This glass insert is designed for applications with limited sample volume. It offers 250 μL in volume and sits securely within a vial. Designed to work with autosamplers, it can be used with wide or standard opening vials. A separately sold spring is required to center it and prevent contact with vial walls.
  • 250 μL volume.
  • Securely fits within vials.
  • Compatible with autosamplers.
  • Usable with wide or standard opening vials.
  • Ensures cleanliness and dimensional tolerances through strict QC.
  • Each unit inspected and tested prior to shipping.
  • Ideal for limited sample volumes.
  • Requires spring to prevent misalignment for sample injection.
Catalog No. 50-785-887 Supplier Waters Corp Supplier No. WAT072704
This glass insert is designed for applications with limited sample volume. It offers 250 μL in volume and sits securely within a vial. Designed to work with autosamplers, it can be used with wide or standard opening vials. A separately sold spring is required to center it and prevent contact with vial walls.
  • 250 μL volume.
  • Securely fits within vials.
  • Compatible with autosamplers.
  • Usable with wide or standard opening vials.
  • Ensures cleanliness and dimensional tolerances through strict QC.
  • Each unit inspected and tested prior to shipping.
  • Ideal for limited sample volumes.
  • Requires spring to prevent misalignment for sample injection.