Learn More
Waters Corp Low Volume Plastic Insert, 250 µL Volume, 144/pk
Catalog No. 50785883 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Vial inserts are a solution in applications where there is limited sample volume. Choose the appropriate volume insert for your sample volume and vial size.
Catalog No. 50-785-883 Supplier Waters Corp Supplier No. WAT072030
Vial inserts are a solution in applications where there is limited sample volume. Choose the appropriate volume insert for your sample volume and vial size.