Learn More
Instech Low-volume luer injection port. Fits into standard female luer hubs, converting a luer into a PinPort and reducing dead volume significantly. Access with PNP3M injector. Sterile.
SDP
Catalog No. NC1612002
Encompass
The Instech LVP3F is a specialty low-volume injection port designed to convert the female luer of certain IV catheters into a PinPort™, displacing the dead volume for critical applications. It provides closed-system benefits while ensuring the luer volume is fully displaced. The port is specifically compatible with the Terumo SurFlash Safety IV catheter; while it may fit other luers, it might bottom out before sealing. Users can access the LVP3F manually with a PinPort™ injector or use the mating connector for continuous tethered infusion.
Features
  • Type: low-volume luer injection port
  • Compatibility: 25ga connectors and Terumo SurFlash catheters
  • Functions: converts luer to PinPort™; reduces dead volume
  • Application: critical dosing and closed-system access
  • Quantity: sold individually
  • Sterility: supplied sterile
  • Material: medical grade plastics and silicone
Catalog No. NC1612002 Supplier Instech Supplier No. LVP3F
The Instech LVP3F is a specialty low-volume injection port designed to convert the female luer of certain IV catheters into a PinPort™, displacing the dead volume for critical applications. It provides closed-system benefits while ensuring the luer volume is fully displaced. The port is specifically compatible with the Terumo SurFlash Safety IV catheter; while it may fit other luers, it might bottom out before sealing. Users can access the LVP3F manually with a PinPort™ injector or use the mating connector for continuous tethered infusion.
Features
  • Type: low-volume luer injection port
  • Compatibility: 25ga connectors and Terumo SurFlash catheters
  • Functions: converts luer to PinPort™; reduces dead volume
  • Application: critical dosing and closed-system access
  • Quantity: sold individually
  • Sterility: supplied sterile
  • Material: medical grade plastics and silicone