Learn More
MilliporeSigma™ Supelco™ Low Volume Static Mixer (50 to 250 μL) Housing, Binary
Catalog No. 111003701 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Material:
PEEK
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
11-100-3701 Stainless Steel
11-100-3599 PEEK
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-3701 Supplier MilliporeSigma™ Supelco™ Supplier No. 57549
Specifications
Material Stainless Steel
Description Fitting: 10-32 ASI in-line housing for 1/16 in. O.D. tubing
For Use With (Equipment) ASI Low Volume Mixer Cartridge
Volume (Metric) 50 to 250 μL