Learn More
LAB BIOREAGENTS LowBinding IV Buffer 500ml
Catalog No. 501959264
Encompass
Lab Bioreagents LowBindings (I, II, III, IV, V, VI, VII) are newly developed assay buffers that provides exceptional performance and broad compatibility for Western blot, ELISA, IHC and nucleic acid detection assays. The LowBindings helps to reduce the non-specific bindings, negative effects of interfering molecules, and cross-reactivities of antibodies and proteins from biomatrix. Therefore, LowBindings exhibit the low matrix interference and high signal-to-noise, by which the assay sensitivity and specificity have been significantly improved. As many factors are involved in immunoassays, there is no single assay buffer that is ideal for each assay. LowBindings (I, II, III, IV, V, VI, VII) are a variety of buffers based on different formulations.
Catalog No. 50-195-9264 Supplier LAB BIOREAGENTS Supplier No. LBA0040500
Lab Bioreagents LowBindings (I, II, III, IV, V, VI, VII) are newly developed assay buffers that provides exceptional performance and broad compatibility for Western blot, ELISA, IHC and nucleic acid detection assays. The LowBindings helps to reduce the non-specific bindings, negative effects of interfering molecules, and cross-reactivities of antibodies and proteins from biomatrix. Therefore, LowBindings exhibit the low matrix interference and high signal-to-noise, by which the assay sensitivity and specificity have been significantly improved. As many factors are involved in immunoassays, there is no single assay buffer that is ideal for each assay. LowBindings (I, II, III, IV, V, VI, VII) are a variety of buffers based on different formulations.