Learn More
Chemglass Life Sciences Lower Reactor Support for Dual Stand, 2 and 5L Vessel Capacity, PTFE Coated
SDP
Catalog No. 501531296
Encompass_Preferred
Lower Reactor Support for Dual Stand, 2 and 5L Vessel Capacity, PTFE Coated. Supports TWO Reactors side by side from 300mL to 5L. Pictured is the full CG-1949-X-20 support stand.
Catalog No. 50-153-1296 Supplier Chemglass Life Sciences Supplier No. CG1949X23
Lower Reactor Support for Dual Stand, 2 and 5L Vessel Capacity, PTFE Coated. Supports TWO Reactors side by side from 300mL to 5L. Pictured is the full CG-1949-X-20 support stand.