Learn More
Chemglass Life Sciences 3L Lower Reservoir
SDP
Catalog No. 501531249
Encompass_Preferred
3L Lower Reservoir. 3L 5-Neck heavy wall, round bottom flask lower reservoir for gas scrubber. Flask has a 60mm Duran Flange center neck, 2ea 45/50 outer joints, and 2ea 24/40 outer joints. Drain valve is a 0-14mm with Perfluoro o-rings. Valve is offset from the bottom of the flask to permit solids to settle and not clog valve or damage the fluid pump head. Connection on the outlet of the valve is 1/2 Swagelok glass tubing for use with CG-1975 compression fitting.
Catalog No. 50-153-1249 Supplier Chemglass Life Sciences Supplier No. CG183012
3L Lower Reservoir. 3L 5-Neck heavy wall, round bottom flask lower reservoir for gas scrubber. Flask has a 60mm Duran Flange center neck, 2ea 45/50 outer joints, and 2ea 24/40 outer joints. Drain valve is a 0-14mm with Perfluoro o-rings. Valve is offset from the bottom of the flask to permit solids to settle and not clog valve or damage the fluid pump head. Connection on the outlet of the valve is 1/2 Swagelok glass tubing for use with CG-1975 compression fitting.