Learn More
Waters Corp Needlewash Frit, 5/pk
Supplier:  Waters Corp 700001318
Encompass_Preferred
Replacement needlewash frit. This is the lower seal wash frit for the needlewash that sits between the lower seal wash body and the filter retainer. There are 5 frits in this package. Frit is also part of Seal Pack Rebuild kit WAT271019.
Catalog No. 50-818-918