Learn More
Chemglass Life Sciences Lower Support Ring for Reaction Vessel, PTFE Coated Steel, 100mm ID, 44mm Slot Opening
SDP
Catalog No. CG194802
Encompass_Preferred
Lower Support Ring for Reaction Vessel, PTFE Coated Stainless Steel, 100mm ID, 44mm Slot Opening. Support ring is constructed of 1/2" OD. PTFE coated stainless steel rod. Made with an opening for easy insertion of reactor with an 8in long rod for connection to lattice.
Catalog No. CG194802 Supplier Chemglass Life Sciences Supplier No. CG194802
Lower Support Ring for Reaction Vessel, PTFE Coated Stainless Steel, 100mm ID, 44mm Slot Opening. Support ring is constructed of 1/2" OD. PTFE coated stainless steel rod. Made with an opening for easy insertion of reactor with an 8in long rod for connection to lattice.