Learn More
Chemglass Life Sciences Lower Support Ring for Reaction Vessel, PTFE Coated Steel, 127mm ID, 79mm Slot Opening
SDP
Catalog No. CG194803
Encompass_Preferred
Lower Support Ring for Reaction Vessel, PTFE Coated Stainless Steel, 127mm ID, 79mm Slot Opening. Support ring is constructed of 1/2" OD. PTFE coated stainless steel rod. Made with an opening for easy insertion of reactor with an 8in long rod for connection to lattice.
Catalog No. CG194803 Supplier Chemglass Life Sciences Supplier No. CG194803
Lower Support Ring for Reaction Vessel, PTFE Coated Stainless Steel, 127mm ID, 79mm Slot Opening. Support ring is constructed of 1/2" OD. PTFE coated stainless steel rod. Made with an opening for easy insertion of reactor with an 8in long rod for connection to lattice.