Learn More
BURKLE INC Lowering cable V4A, length 32.81 ft. (10m), Ø 0.05 in. (1,25 mm)
Catalog No. 5000025861
Encompass_Preferred
The 316L descent cable delivers outstanding chemical resistance and durability, making it perfect for demanding sampling applications. Designed for use with Burkle dipping vessels, diving cylinders, and diving bombs, it handles a maximum tensile load of up to 10 kg (22.05 lbs) with ease. Its robust, corrosion-resistant construction ensures reliable performance even in hazardous areas and aggressive chemical environments.
Catalog No. 50-000-25861 Supplier BURKLE INC Supplier No. 53090301
The 316L descent cable delivers outstanding chemical resistance and durability, making it perfect for demanding sampling applications. Designed for use with Burkle dipping vessels, diving cylinders, and diving bombs, it handles a maximum tensile load of up to 10 kg (22.05 lbs) with ease. Its robust, corrosion-resistant construction ensures reliable performance even in hazardous areas and aggressive chemical environments.