Learn More
BURKLE INC Copper lowering cable, length 164.04 ft. (50m), Ø 0.18 in. (4,5 mm)
Catalog No. 5000025550
Encompass_Preferred
The EX copper rope with loops is specially designed for use in potentially explosive atmospheres. Made from high quality copper, a non-sparking metal, it offers maximum safety when used in potentially explosive atmospheres. The loops allow for easy attachment to sampling cylinders and bombs and fit perfectly with hand rope reels made of various materials. The maximum tensile load is 10 kg (22.05 pd.).
Catalog No. 50-000-25550 Supplier BURKLE INC Supplier No. 53090450
The EX copper rope with loops is specially designed for use in potentially explosive atmospheres. Made from high quality copper, a non-sparking metal, it offers maximum safety when used in potentially explosive atmospheres. The loops allow for easy attachment to sampling cylinders and bombs and fit perfectly with hand rope reels made of various materials. The maximum tensile load is 10 kg (22.05 pd.).