Learn More
BURKLE INC Lowering cable V2A/PTFE, length 82.02 ft. (25m), Ø 0.08 in. (2 mm)
Catalog No. 5000025631
Encompass_Preferred
The 304/PTFE descender rope is characterized by its high purity materials. The PTFE coating prevents the rope from getting wet and allows liquids to run off without leaving a residue. With a tensile load of up to 10 kg (22.05 pd.), it is ideal for use with immersion samplers such as diving bottles and diving bombs. Excellent chemical resistance ensures a long service life and flexible, reliable operation.
Catalog No. 50-000-25631 Supplier BURKLE INC Supplier No. 53090125
The 304/PTFE descender rope is characterized by its high purity materials. The PTFE coating prevents the rope from getting wet and allows liquids to run off without leaving a residue. With a tensile load of up to 10 kg (22.05 pd.), it is ideal for use with immersion samplers such as diving bottles and diving bombs. Excellent chemical resistance ensures a long service life and flexible, reliable operation.