Learn More
BURKLE INC Lowering cable PP, length 32.81 ft. (10m), Ø 0.08 in. (2 mm)
Catalog No. 5000025683
Encompass_Preferred
Made from durable red PP cord, the Plastic Lowering Cable provides a cost-effective solution for use with dip bottles and immersion samplers. With a maximum tensile load of up to 10 kg (22.05 pd.), it is robust and reliable. Suitable for hand reels made from a variety of materials, it offers a wide range of applications.
Catalog No. 50-000-25683 Supplier BURKLE INC Supplier No. 53090201
Made from durable red PP cord, the Plastic Lowering Cable provides a cost-effective solution for use with dip bottles and immersion samplers. With a maximum tensile load of up to 10 kg (22.05 pd.), it is robust and reliable. Suitable for hand reels made from a variety of materials, it offers a wide range of applications.