Learn More
BURKLE INC Stainless steel chain V4A, length 32.81 ft. (10m), Ø0.09 in. (2,3 mm)
Catalog No. 5000025936
Encompass_Preferred
The drop/pull chain is a high quality 316TI stainless steel (1.4571) chain suitable for both drop and pull applications. Its excellent corrosion resistance and durability make it ideal for use in marine, construction, industrial and various outdoor applications. It is robust, reliable and versatile.
Catalog No. 50-000-25936 Supplier BURKLE INC Supplier No. 53090510
The drop/pull chain is a high quality 316TI stainless steel (1.4571) chain suitable for both drop and pull applications. Its excellent corrosion resistance and durability make it ideal for use in marine, construction, industrial and various outdoor applications. It is robust, reliable and versatile.