Learn More
Electron Microscopy Sciences Lowicryl K11M Kit
SDP
Catalog No. 50980415
Encompass
Lowicryl resins are highly cross-linked acrylate-based embedding resins, designed for use over a wide range of embedding conditions. These resins provide low viscosity at low temperatures. All of these resins are photopolymerized by long wavelength (360nm) ultraviolet light. Since the initiation of the polymerization is largely independent of temperatures, blocks may be polymerized at the same temperatures used for infiltration. The resin may also be chemically polymerized at 60 degree C. Usable to -60 deg C.
Catalog No. 50-980-415 Supplier Electron Microscopy Sciences Supplier No. 14350
Lowicryl resins are highly cross-linked acrylate-based embedding resins, designed for use over a wide range of embedding conditions. These resins provide low viscosity at low temperatures. All of these resins are photopolymerized by long wavelength (360nm) ultraviolet light. Since the initiation of the polymerization is largely independent of temperatures, blocks may be polymerized at the same temperatures used for infiltration. The resin may also be chemically polymerized at 60 degree C. Usable to -60 deg C.