Learn More
Electron Microscopy Sciences Lowicryl K4M Monostep 225 GR
SDP
Catalog No. 50980412
Encompass
The same as our Lowicryl complete kits but this new formulation allows you one stop embedding without all of the cumbersome preparation and mixing.  This pre-mixed, ready to use resin, saves you time and minimizes chemical contact.  MonoStep Lowicryl is ideal for use in immunohistochemistry and immunolabeling.  Store at room temperature.
Catalog No. 50-980-412 Supplier Electron Microscopy Sciences Supplier No. 14335
The same as our Lowicryl complete kits but this new formulation allows you one stop embedding without all of the cumbersome preparation and mixing.  This pre-mixed, ready to use resin, saves you time and minimizes chemical contact.  MonoStep Lowicryl is ideal for use in immunohistochemistry and immunolabeling.  Store at room temperature.