Learn More
Labcon North America Labcon ZAP SLIK 1000 µL Aerosol Filter Tips, Low Retention, Sterile, Wide Orifice
Catalog No. NC2746974
Encompass_Preferred
The Labcon ZAP™ SLIK 1000 µL Low Retention Aerosol Filter Pipette Tips are designed for precise liquid handling, enhancing sample recovery with low retention technology. The wide orifice allows for gentle transfer of fragile cells like embryonic stem cells. Rigorously tested for purity, these sterile tips prevent contamination and protect both samples and pipettors, ensuring reliable results in sensitive applications. Made in the USA, they uphold high environmental standards and are compatible with most pipette brands.
Catalog No. NC2746974 Supplier Labcon North America Supplier No. 11489650089
The Labcon ZAP™ SLIK 1000 µL Low Retention Aerosol Filter Pipette Tips are designed for precise liquid handling, enhancing sample recovery with low retention technology. The wide orifice allows for gentle transfer of fragile cells like embryonic stem cells. Rigorously tested for purity, these sterile tips prevent contamination and protect both samples and pipettors, ensuring reliable results in sensitive applications. Made in the USA, they uphold high environmental standards and are compatible with most pipette brands.