Learn More
Medchemexpress LLC LOX-1/OLR1 Human H 100ug
SDP
Catalog No. 5000197196
Encompass_Preferred
Recombinant human LOX-1/OLR1 ectodomain (S61-Q273) produced in HEK293 cells and supplied lyophilized with a C-terminal His tag. The preparation is >95% pure and formulated for stability, enabling biochemical and binding assays that require a properly folded, glycosylated human receptor fragment.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Includes C-terminal 6xHis tag for affinity detection and purification.
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.2 for improved stability.
  • Store lyophilized at -20 °C; aliquot and freeze at -80 °C after reconstitution for long-term storage.
  • Suitable for binding assays, biochemical characterization, and assay development.
Catalog No. 50-001-97196 Supplier Medchemexpress LLC Supplier No. HYP71180100UG
Recombinant human LOX-1/OLR1 ectodomain (S61-Q273) produced in HEK293 cells and supplied lyophilized with a C-terminal His tag. The preparation is >95% pure and formulated for stability, enabling biochemical and binding assays that require a properly folded, glycosylated human receptor fragment.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Includes C-terminal 6xHis tag for affinity detection and purification.
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.2 for improved stability.
  • Store lyophilized at -20 °C; aliquot and freeze at -80 °C after reconstitution for long-term storage.
  • Suitable for binding assays, biochemical characterization, and assay development.