Learn More
Medchemexpress LLC LOX-1/OLR1 Human H 1mg
SDP
Catalog No. 5000196453
Encompass_Preferred
Recombinant human LOX-1 (OLR1) protein supplied as a lyophilized, C-terminal His-tagged fragment produced in mammalian cells. This reagent is intended for biochemical assays, ligand-binding studies, antibody validation, and other research applications requiring purified LOX-1 with native-like post-translational modifications.
  • Expressed in HEK293 cells for mammalian post-translational modifications.
  • C-terminal His tag for facile purification and detection.
  • Lyophilized formulation for long-term storage and convenient reconstitution.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Recommended minimum reconstitution concentration: 100 μg/mL.
  • Suitable for binding assays, antibody development, and biochemical characterization.
  • Contains the receptor fragment corresponding to S61-Q273 of the human sequence.
Catalog No. 50-001-96453 Supplier Medchemexpress LLC Supplier No. HYP711801MG
Recombinant human LOX-1 (OLR1) protein supplied as a lyophilized, C-terminal His-tagged fragment produced in mammalian cells. This reagent is intended for biochemical assays, ligand-binding studies, antibody validation, and other research applications requiring purified LOX-1 with native-like post-translational modifications.
  • Expressed in HEK293 cells for mammalian post-translational modifications.
  • C-terminal His tag for facile purification and detection.
  • Lyophilized formulation for long-term storage and convenient reconstitution.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Recommended minimum reconstitution concentration: 100 μg/mL.
  • Suitable for binding assays, antibody development, and biochemical characterization.
  • Contains the receptor fragment corresponding to S61-Q273 of the human sequence.