Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC LOX-1/OLR1 Human H 500ug
SDP

Catalog No. 5000196530
Encompass_Preferred

Recombinant human LOX-1/OLR1 extracellular domain expressed in HEK293 cells and supplied lyophilized with a C-terminal 6xHis tag. The protein is highly pure and formulated for reliable reconstitution and storage for research applications.

  • Recombinant extracellular domain (S61-Q273).
  • Expressed in HEK293 cells.
  • C-terminal 6xHis tag for purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.2.
  • Recommended reconstitution concentration of at least 100 μg/mL in ddH2O.
  • Store at -20°C; aliquot and freeze at -80°C for extended storage.
  • For research use only.

Catalog No. 50-001-96530 Supplier Medchemexpress LLC Supplier No. HYP71180500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human LOX-1/OLR1 extracellular domain expressed in HEK293 cells and supplied lyophilized with a C-terminal 6xHis tag. The protein is highly pure and formulated for reliable reconstitution and storage for research applications.

  • Recombinant extracellular domain (S61-Q273).
  • Expressed in HEK293 cells.
  • C-terminal 6xHis tag for purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.2.
  • Recommended reconstitution concentration of at least 100 μg/mL in ddH2O.
  • Store at -20°C; aliquot and freeze at -80°C for extended storage.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.