Learn More
Medchemexpress LLC LOX-1/OLR1 Human H 5ug
SDP
Catalog No. 5000197232
Encompass_Preferred
Recombinant human LOX-1/OLR1 extracellular domain expressed in HEK293 cells and supplied lyophilized with a C-terminal His tag. The protein is >95% pure by reducing SDS-PAGE, formulated in phosphate buffer with sodium chloride, and provided in a range of microgram to milligram sizes. Store lyophilized at -20°C; after reconstitution store short term at 4°C or aliquot and freeze at -20°C/-80°C for long-term storage. For research use only.
  • Recombinant extracellular domain suitable for binding and biochemical assays.
  • Expressed in mammalian cells with a C-terminal 6xHis tag to enable affinity purification.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.2 for stability.
  • Reconstitute at ≥100 μg/mL in ddH2O; aliquot and freeze for long-term storage.
  • Intended for research use only.
Catalog No. 50-001-97232 Supplier Medchemexpress LLC Supplier No. HYP711805UG
Recombinant human LOX-1/OLR1 extracellular domain expressed in HEK293 cells and supplied lyophilized with a C-terminal His tag. The protein is >95% pure by reducing SDS-PAGE, formulated in phosphate buffer with sodium chloride, and provided in a range of microgram to milligram sizes. Store lyophilized at -20°C; after reconstitution store short term at 4°C or aliquot and freeze at -20°C/-80°C for long-term storage. For research use only.
  • Recombinant extracellular domain suitable for binding and biochemical assays.
  • Expressed in mammalian cells with a C-terminal 6xHis tag to enable affinity purification.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Lyophilized from 20 mM PB, 150 mM NaCl, pH 7.2 for stability.
  • Reconstitute at ≥100 μg/mL in ddH2O; aliquot and freeze for long-term storage.
  • Intended for research use only.