Learn More
Medchemexpress LLC LOX-1/OLR1 Rat HEK 100ug
SDP
Catalog No. 5000179006
Encompass_Preferred
The LOX-1/OLR1 protein plays an important role as a receptor that recognizes internalizes and degrades oxidatively modified low-density lipoprotein (oxLDL) a marker of atherosclerosis in vascular endothelial cells OxLDL induces endothelial dysfunction triggering pro-inflammatory responses oxidative conditions and apoptosis LOX-1/OLR1 Protein Rat (HEK293 His) is the recombinant rat-derived LOX-1/OLR1 protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-79006 Supplier Medchemexpress LLC Supplier No. HYP73835100UG
The LOX-1/OLR1 protein plays an important role as a receptor that recognizes internalizes and degrades oxidatively modified low-density lipoprotein (oxLDL) a marker of atherosclerosis in vascular endothelial cells OxLDL induces endothelial dysfunction triggering pro-inflammatory responses oxidative conditions and apoptosis LOX-1/OLR1 Protein Rat (HEK293 His) is the recombinant rat-derived LOX-1/OLR1 protein expressed by HEK293 with N-His labeled tag